Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04290585

Isorhamnetin

primary reference standard

Synonym(s):

3′-Methoxy-3,4′,5,7-tetrahydroxyflavone, 3′-Methylquercetin, 3,4′,5,7-Tetrahydroxy 3′-methoxyflavone, Isorhamnetol, Quercetin 3′-methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O7
CAS Number:
Molecular Weight:
316.26
Beilstein/REAXYS Number:
44723
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

SMILES string

COc1cc(ccc1O)C2=C(O)C(=O)c3c(O)cc(O)cc3O2

InChI

1S/C16H12O7/c1-22-11-4-7(2-3-9(11)18)16-15(21)14(20)13-10(19)5-8(17)6-12(13)23-16/h2-6,17-19,21H,1H3

InChI key

IZQSVPBOUDKVDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Biochem/physiol Actions

Isorhamnetin inhibits adipogenesis by interfering with differentiation of adipose stem cells, by a mechanism involving stabilization of β-catenin and up-regulating the Wnt signaling pathway.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong-Woon Cho et al.
Archives of pharmacal research, 37(9), 1159-1168 (2014-01-22)
Fermentation of natural products is emerging as an important processing method and is attracting a lot of attention because it may have the advantage of having a new biological function. In this study, fruits of Opuntia ficus-indica were enzymatically hydrolyzed
Joseph A Lutz et al.
Fitoterapia, 98, 11-21 (2014-06-28)
The alpha7 nicotinic acetylcholine receptor (nAChR) is a potential target in neuroinflammation. Screening a plant extract library identified Solidago nemoralis as containing methyl-quercetin derivatives that are relatively selective ligands for the alpha7 nAChR. Flavonoids are not known for this activity
Claudine Valérie Passo Tsamo et al.
Food chemistry, 167, 197-204 (2014-08-26)
The present study investigated the phenolic profiles of the pulp and peel of nine plantain cultivars and compared them to those of two dessert bananas of commercial interest (Grand Nain and Gros Michel), alongside a newly created hybrid, resistant to
Haixia Lin et al.
Toxicological sciences : an official journal of the Society of Toxicology, 139(2), 338-349 (2014-03-07)
Ginkgo biloba has been used for many thousand years as a traditional herbal remedy and its extract has been consumed for many decades as a dietary supplement. Ginkgo biloba leaf extract is a complex mixture with many constituents, including flavonol
Bao-Yi Zhang et al.
International journal of clinical and experimental pathology, 8(1), 25-37 (2015-03-11)
The development of novel antitumor drugs for the treatment of non-small cell lung carcinoma NSCLC is imperative in order to improve the efficacy of lung cancer therapy and prognosis. In the current study, we demonstrated the antitumor activity of isorhamnetin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service