04175
Aldicarb-(N-methyl-13C,d3, carbamoyl-13C) sulfoxide
PESTANAL®, analytical standard
About This Item
grade
analytical standard
Quality Level
isotopic purity
≥99.0 atom % 13C
≥98.0 atom % D
product line
PESTANAL®
assay
≥98.5% (HPLC)
shelf life
limited shelf life, expiry date on the label
application(s)
agriculture
environmental
format
neat
mass shift
M+5
storage temp.
−20°C
−20°C
SMILES string
[2H][13C]([2H])([2H])N[13C](=O)O\N=C\C(C)(C)S(C)=O
InChI
1S/C7H14N2O3S/c1-7(2,13(4)11)5-9-12-6(10)8-3/h5H,1-4H3,(H,8,10)/b9-5+/i3+1D3,6+1
InChI key
BXPMAGSOWXBZHS-FONWFMSDSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 1 Oral - Aquatic Acute 1
Storage Class
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service