Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04174

Supelco

1-Acetonaphthone

analytical standard

Synonym(s):

1-Acetylnaphthalene, Methyl 1-naphthyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7COCH3
CAS Number:
Molecular Weight:
170.21
Beilstein/REAXYS Number:
1100618
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

assay

≥97.0% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

refractive index

n20/D 1.627-1.629
n20/D 1.628 (lit.)

bp

302 °C (lit.)

mp

10.5 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

SMILES string

CC(=O)c1cccc2ccccc12

InChI

1S/C12H10O/c1-9(13)11-8-4-6-10-5-2-3-7-12(10)11/h2-8H,1H3

InChI key

QQLIGMASAVJVON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

>212.0 °F

flash_point_c

> 100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Babu Balaji et al.
European journal of medicinal chemistry, 92, 332-341 (2015-01-13)
Oxovanadium(IV) complexes [VO(Fc-tpy)(acac)](ClO4) (1), [VO(Fc-tpy)(nap-acac)](ClO4) (2), [VO(Fc-tpy)(py-acac)](ClO4) (3) and [VO(Ph-tpy)(py-acac)](ClO4) (4) of 4'-ferrocenyl-2,2':6',2"-terpyridine (Fc-tpy) and 4'-phenyl-2,2':6',2"-terpyridine (Ph-tpy) having monoanionic acetylacetonate (acac), naphthylacetylacetonate (nap-acac) or pyrenylacetylacetonate (py-acac) ligand were prepared, characterized and their photocytotoxicity in visible light studied. The ferrocenyl complexes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service