Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04124

Sigma-Aldrich

1-(3-Cyanopropyl)-3-methylimidazolium chloride

Task Specific Ionic Liquid (TSIL), ≥98.5% (T)

Synonym(s):

1-Methyl-3-(3-cyanopropyl)imidazolium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12ClN3
CAS Number:
Molecular Weight:
185.65
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥98.5% (T)
≥98.5% (elemental analysis, N)

form

powder

impurities

≤0.5% water

SMILES string

[Cl-].C[n+]1ccn(CCCC#N)c1

InChI

1S/C8H12N3.ClH/c1-10-6-7-11(8-10)5-3-2-4-9;/h6-8H,2-3,5H2,1H3;1H/q+1;/p-1

InChI key

HGPSWUJKMXIBKA-UHFFFAOYSA-M

General description

Nitrile-functionalized ionic liquid showing superior characteristics in Suzuki and Stille coupling reactions[1].

Application

1-(3-Cyanopropyl)-3-methylimidazolium chloride can be used:
  • To prepare ionic polymers stabilized with palladium nanoparticle catalysts, which are applicable in the hydrogenation of alkenes and arenes.[2]
  • As a solvent in the synthesis of substituted 4-arylthio-1,2-oxazoles by reacting different aryl iodides with acetylenic oximes in the presence of Pd-NHC catalyst.[3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances in sulfuration chemistry enabled by bunte salts
Wang M, et al.
Aldrichimica Acta, 53(1), 19-19 (2020)
Aqueous-phase hydrogenation of alkenes and arenes: The growing role of nanoscale catalysts
Bulut S, et al.
Catalysis Today, 247, 96-103 (2015)
Dongbin Zhao et al.
Journal of the American Chemical Society, 126(48), 15876-15882 (2004-12-02)
A series of relatively low-cost ionic liquids, based on the N-butyronitrile pyridinium cation [C(3)CNpy](+), designed to improve catalyst retention, have been prepared and evaluated in Suzuki and Stille coupling reactions. Depending on the nature of the anion, these salts react

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service