Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04029

Supelco

4-Nonylphenol

certified reference material, TraceCERT®

Synonym(s):

4-n-Nonylphenol, p-n-Nonylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
Beilstein/REAXYS Number:
2047450
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

technique(s)

gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCc1ccc(O)cc1

InChI

1S/C15H24O/c1-2-3-4-5-6-7-8-9-14-10-12-15(16)13-11-14/h10-13,16H,2-9H2,1H3

InChI key

IGFHQQFPSIBGKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com.

4-Nonylphenol (NP) belongs to the class of phenolic xenoestrogens, with estrogen effects, which adversely affect the health and reproduction of humans and animals.[1] It is released into the environment as a degradation product of alkylphenol ethoxylates (APEOs), widely used in industrial, household, and commercial applications.[2] This compound is also used as a precursor in the production of non-ionic surfactants.[1]

Application

4-Nonylphenol may be used as an analytical reference standard for the quantification of the analyte in environmental samples,[1] fish and shellfish,[2] biological samples[3] using chromatography techniques.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 4-tert-octylphenol, 4-nonylphenol and bisphenol A in surface waters from the Haihe River in Tianjin by gas chromatography-mass spectrometry with selected ion monitoring.
Jin X, et al.
Chemosphere, 56(11), 1113-1119 (2004)
Determination of 4-nonylphenol, nonylphenol monoethoxylate, nonylphenol diethoxylate and other alkylphenols in fish and shellfish by high-performance liquid chromatography with fluorescence detection.
Tsuda T, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 746(2), 305-309 (2000)
Determination of 4-nonylphenol and 4-tert.-octylphenol in water samples by stir bar sorptive extraction and thermal desorption-gas chromatography-mass spectrometry.
Kawaguchi M, et al.
Analytica Chimica Acta, 505(2), 217-222 (2004)

Articles

Nonylphenols (NP) are ubiquitous substances that have been detected in highly diverse foodstuff. As breakdown products of nonylphenol ethoxylates, NP could end up in the aquatic environment. This article presents a rapid method for NP detection in surface water by planar solid phase extraction – planar yeast estrogen screen (pSPE-pYES).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service