Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

04016

Supelco

3-Pentadecylphenol

analytical standard

Synonym(s):

2-Deoxy-urushiol I

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14C6H4OH
CAS Number:
Molecular Weight:
304.51
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

composition

carbon content, 82.83%
hydrogen content, 11.92%

bp

190-195 °C/1 mmHg (lit.)

mp

50-53 °C (lit.)
52-55 °C

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCc1cccc(O)c1

InChI

1S/C21H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-20-17-15-18-21(22)19-20/h15,17-19,22H,2-14,16H2,1H3

InChI key

PTFIPECGHSYQNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Pentadecylphenol is classified under the amphiphilic phenolic lipids group of chemicals.[1]

Application

3-Pentadecylphenol may be used as a reference standard for the analysis of 3-pentadecylphenol as a bioactive compound in Aplotaxis auriculata rhizome by gas chromatography-mass spectrometry (GC-MS).[2] It has been reported to have been used as a reference standard for the determination of 3-pentadecylphenol in Sakhalin-Ainu[3] as well as Ryukyu[4] lacquerwares and in Japanese ancient lacquerwares excavated from Jomon period ruins[5] by pyrolysis GC-MS.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of Ryukyu lacquerwares by pyrolysis-gas chromatography/mass spectrometry.
Honda, et al.
Journal of Analytical and Applied Pyrolysis, 113(1), 41-45 (2015)
Analysis of Sakhalin-Ainu lacquerwares by pyrolysis gas chromatography/mass spectrometry.
Idei, et al.
Journal of archaeological science, reports., 20(1), 1-5 (2018)
The effect of 3-pentadecylphenol on DPPC bilayers ATR-IR and 31P NMR studies.
Cieslik-Boczula, et al.
Biophysical Chemistry, 140(1-3), 51-56 (2009)
Analysis of Japanese ancient lacquerwares excavated from J?mon period ruins.
Yuasa, et al.
Journal of Analytical and Applied Pyrolysis, 113(1), 73-77 (2015)
Determination of bioactive compounds in Aplotaxis auriculata DC. rhizome extract using GC-MS technique.
Shalini, et al.
Asian journal of innovative Research, 2(1), 49-53 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service