Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03936

Supelco

Betamethasone 21-valerate

pharmaceutical impurity standard

Synonym(s):

Betamethasone Related Compound A (USP), Betamethasone impurity E (PhEur)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H37FO6
CAS Number:
Molecular Weight:
476.58
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤10.0% related substances
≤5.0% solvents
≤5.0% water

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

SMILES string

CCCCC(=O)OCC(=O)[C@@]1(O)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C

InChI

1S/C27H37FO6/c1-5-6-7-23(32)34-15-22(31)27(33)16(2)12-20-19-9-8-17-13-18(29)10-11-24(17,3)26(19,28)21(30)14-25(20,27)4/h10-11,13,16,19-21,30,33H,5-9,12,14-15H2,1-4H3/t16-,19-,20-,21-,24-,25-,26-,27-/m0/s1

InChI key

FEROCCAEIIKMJT-SUYDQAKGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Betamethasone 21-valerate is an active pharmaceutical ingredient present in various topical pharmaceutical creams.[1]

Application

Betamethasone 21-valerate may be used as a reference standard in the determination of betamethasone 21-valerate in counterfeit drugs using high-performance liquid chromatography coupled with electrospray ionization mass spectrometry (HPLC/ESI‐MS).[2] It may be used as a pharmaceutical impurity standard in the quantification of impurities of betamethasone 21-valerate in topical pharmaceutical creams using high-performance liquid chromatography (HPLC).[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An RP-HPLC method for the stability-indicating analysis of impurities of both fusidic acid and betamethasone-17-valerate in a semi-solid pharmaceutical dosage form.
Byrne J, et al.
Journal of Chromatographic Science, 53(9), 1498-1503 (2015)
Analysis of betamethasone, dexamethasone and related compounds by liquid chromatography/electrospray mass spectrometry.
Arthur EK, et al.
Rapid Communications in Mass Spectrometry, 18(6), 678-684 (2004)
K Kubota et al.
Journal of pharmaceutical sciences, 84(12), 1478-1481 (1995-12-01)
Simultaneous diffusion and metabolism of betamethasone 17-valerate was studied using betamethasone 17-valerate, betamethasone 21-valerate, and betamethasone as permeants. These corticosteroids were suspended in silicone adhesive and applied to an artificial living skin equivalent (LSE) for 72 h. When betamethasone was
R De Pasquale et al.
Giornale italiano di dermatologia e venereologia : organo ufficiale, Societa italiana di dermatologia e sifilografia, 145(1), 135-138 (2010-03-04)
Lichen amyloidosis is a primary localized cutaneous amyloidosis without systemic involvement, characterized by a persistent pruritic eruption of multiple discrete hyperkeratotic papules. The etiology is unknown, but chronic irritation of the skin has been proposed as an etiological factor. We
K Kubota et al.
Dermatology (Basel, Switzerland), 188(1), 13-17 (1994-01-01)
The metabolism of betamethasone 17-valerate was estimated using an artificial living skin equivalent (LSE). Betamethasone 17-valerate, betamethasone 21-valerate and betamethasone were measured by a normal-phase high-performance liquid chromatographic (HPLC) method. Betamethasone 17-valerate was added to the culture medium with or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service