Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03853

Sigma-Aldrich

4-Nonylphenyl-polyethylene glycol

non-ionic

Synonym(s):

(Nonylphenoxy)polyethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

Quality Level

mol wt

average mol wt 680

CMC

0.059 mM (20-25°C)

transition temp

cloud point 45-50 °C

InChI

1S/C19H32O3/c1-2-3-4-5-6-7-8-9-18-10-12-19(13-11-18)22-17-16-21-15-14-20/h10-13,20H,2-9,14-17H2,1H3

InChI key

BLXVTZPGEOGTGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

with approx. 10 ethylene oxide units

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service