Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03796

Supelco

Titanium atomic spectroscopy standard concentrate 1.00 g Ti

1.00 g/L, for 1 l standard solution, analytical standard

Synonym(s):

Titanium(IV) chloride solution, Titanium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl4
CAS Number:
Molecular Weight:
189.68
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

packaging

ampule of

concentration

1.00 g/L

solubility

H2O: soluble

SMILES string

Cl[Ti](Cl)(Cl)Cl

InChI

1S/4ClH.Ti/h4*1H;/q;;;;+4/p-4

Inchi Key

XJDNKRIXUMDJCW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Preparation Note

prepared with TiCl4, HCl and H2O

Reconstitution

Dilute the ampule to approx. 850 mL with water. Carefully add 100 mL conc. HCl to acidify. Bring to volume with water.

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT RE 2 - STOT SE 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bratindranath Mukherjee et al.
Nanoscale, 5(1), 269-274 (2012-11-20)
The surface treatment of an anodized TiO(2) nanotube (T_NT) is very desirable for enhancing its photoelectrochemical properties and often is a prerequisite to deposition of any overlying layer for photoactivity efficiency improvement. This study provides a comparative analysis of the
Yang Chen et al.
ChemSusChem, 4(10), 1498-1503 (2011-09-29)
Mesoporous TiO₂ beads with diameters of 320±50, 550±50, and 830±40 nm, comprising interconnected and densely packed TiO₂ nanocrystals, were used as working electrodes for dye-sensitized solar cells (DSCs). These electrodes possess high surface areas and superior light-scattering properties, which are ideal
Kazuhiko Akiyama et al.
Journal of the American Chemical Society, 134(23), 9762-9767 (2012-05-18)
Rapid and efficient separation/purification of pure metallofullerenes M(x)@C(n) (M = metal; x = 1, 2; n > 70) and carbide metallofullerenes of the type M(y)C(2)@C(n-2) (y = 2, 3, 4; n - 2 > 68) has been reported. The present
Y X Zhao et al.
Bioresource technology, 108, 45-54 (2012-01-31)
The objectives of this study are to investigate the impacts of anionic polymer compound bioflocculant (CBF) as a coagulant aid on coagulation performance and floc characteristics with titanium tetrachloride (TiCl(4)) and aluminum sulfate (Al(2)(SO(4))(3)). The effect of dosing sequence was
Kun Wang et al.
The oncologist, 20(3), 263-269 (2015-02-07)
The TERT promoter and FGFR3 gene mutations are two of the most common genetic events in urothelial bladder cancer (UBC), and these mutation assays in patient urine have been shown to be promising biomarkers for UBC diagnosis and surveillance. These

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service