Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03768

Sigma-Aldrich

′L-β-Homoproline′ hydrochloride

≥98.0% (TLC)

Synonym(s):

(S)-2-(2-Pyrrolidinyl)acetic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO2 · HCl
CAS Number:
Molecular Weight:
165.62
Beilstein/REAXYS Number:
471672
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Quality Level

assay

≥98.0% (TLC)

form

powder

color

white to light brown

application(s)

peptide synthesis

SMILES string

Cl.OC(=O)C[C@@H]1CCCN1

InChI

1S/C6H11NO2.ClH/c8-6(9)4-5-2-1-3-7-5;/h5,7H,1-4H2,(H,8,9);1H/t5-;/m0./s1

InChI key

VQDACVOAOJQTPR-JEDNCBNOSA-N

Application

L-β-homoproline is a non-coded α-imino acid used in organic peptide synthesis.

Biochem/physiol Actions

L-β-Homoproline is a β amino acid useful in peptide and protein conformation studies.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dariusz Sobolewski et al.
Journal of peptide science : an official publication of the European Peptide Society, 15(3), 161-165 (2008-11-28)
Using SPPS techniques, six new analogues of AVP and some of its agonists were synthesised. The peptides were designed by substitution of Phe at position 3 of AVP, [Mpa(1)] AVP (dAVP) and [Mpa(1),Val(4),D-Arg(8)]VP (dVDAVP) with L- or D-Pip, a non-coded
Sándor Góbi et al.
Physical chemistry chemical physics : PCCP, 12(41), 13603-13615 (2010-09-21)
In order to test the pseudo-γ-turn forming capability of β-homo-proline (β(3)-HPro) 2-[(2S)-1-acetylpyrrolidin-2-yl]-N-methylacetamide (Ac-β(3)-HPro-NHMe) was synthesized and its potential energy landscape was investigated by infrared (IR) and vibrational circular dichroism (VCD) spectroscopy combined with density functional calculations. Based upon a comparison
New bradykinin B(2) receptor antagonists - influence of C-terminal segment modifications on their pharmacological properties.
Sleszynska M, Kwiatkowska A, et al.
Acta Biochimica Polonica, 56, 6410648-6410648 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service