Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03382

Supelco

Aluminum atomic spectroscopy standard concentrate 10.00 g Al

10.00 g/L, for 1 l standard solution, analytical standard

Synonym(s):

Aluminum chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AlCl3
CAS Number:
Molecular Weight:
133.34
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

packaging

ampule of

concentration

10.00 g/L

solubility

H2O: soluble

SMILES string

Cl[Al](Cl)Cl

InChI

1S/Al.3ClH/h;3*1H/q+3;;;/p-3

Inchi Key

VSCWAEJMTAWNJL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Preparation Note

prepared with AlCl3.6H2O and H2O

pictograms

Health hazardCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

target_organs

Lungs

supp_hazards

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manal Fouad Ismail et al.
International journal of nanomedicine, 8, 393-406 (2013-02-05)
To sustain the effect of rivastigmine, a hydrophilic cholinesterase inhibitor, nanobased formulations were prepared. The efficacy of the prepared rivastigmine liposomes (RLs) in comparison to rivastigmine solution (RS) was assessed in an aluminium chloride (AlCl(3))-induced Alzheimer's model. Liposomes were prepared
Bagineni Prasad et al.
Chemical communications (Cambridge, England), 48(84), 10434-10436 (2012-09-20)
A conceptually new and straightforward introduction of sulfonyl groups at the C-7 position of an indole ring has been achieved via AlCl(3) mediated unexpected regioselective sulfonyl group migration for N-alkyl/aryl/heteroarylsulfonyl indoles affording potential inhibitors of Mycobacterium tuberculosis H37Rv chorismate mutase.
Diagnosis and management of hyperhidrosis.
R A Benson et al.
BMJ (Clinical research ed.), 347, f6800-f6800 (2013-11-28)
Mengyun Zhu et al.
Organic letters, 16(7), 1856-1859 (2014-03-19)
An unprecedented AlCl3-promoted formal [2 + 3]-cycloaddition of 1,1-cyclopropanes with readily available N-benzylic sulfonamides has been developed. Experimental evidence supports an unusual mechanism wherein the donor-acceptor cyclopropane serves as a source of 2-styrylmalonate rather than the "classical" 1,3-dipole. A broad
Amira Zaky et al.
BMC neuroscience, 14, 26-26 (2013-03-19)
Chronic administration of Aluminum is proposed as an environmental factor that may affect several enzymes and other biomolecules related to neurotoxicity and Alzheimer's disease (AD). APE1 a multifunctional protein, functions in DNA repair and plays a key role in cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service