Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03374

Supelco

Cobalt atomic spectroscopy standard concentrate 10.00 g Co

10.00 g/L, for 1L standard solution, analytical standard

Synonym(s):

Cobalt(II) sulfate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

packaging

ampule of

concentration

10.00 g/L

solubility

H2O: soluble

SMILES string

[Co++].[O-]S([O-])(=O)=O

InChI

1S/Co.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

KTVIXTQDYHMGHF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Preparation Note

prepared with CoSO4.7H2O, H2SO4 and H2O

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Met. Corr. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cobalt sulfate.
Report on carcinogens : carcinogen profiles, 12, 113-115 (2011-08-19)
Carolyn F Moyer et al.
Toxicologic pathology, 30(4), 427-434 (2002-08-22)
Epidemiological studies suggest an association between ambient particulate matter and cardiopulmonary diseases in humans. The mechanisms underlying these health effects are poorly understood. To better understand the potential relationship between particulate-matter-induced inflammation and vascular disease, a 2-phase retrospective study was
S Sahoo et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 26(12), 1309-1312 (2010-12-16)
An analytical method has been described for the determination of mercury using a carbon paste electrode. Conditions for the preparation of the carbon paste electrode were optimized for low background current in order to use it for the determination of
Lei Hou et al.
Chemical communications (Cambridge, England), 46(34), 6311-6313 (2010-08-03)
Solvothermal reaction of CoSO(4) with 2,6-di-p-carboxyphenyl-4,4'-bipyridine affords a novel, octacobalt cluster based, (3,12)-connected porous framework, which exhibits gas sorption and spin-glassy magnetic behaviour.
Environment toxicity as a new universal characteristic of the pollution of objects tested.
P E Tulupov et al.
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections, 399, 496-499 (2005-02-19)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service