Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03372

Supelco

Copper atomic spectroscopy standard concentrate 10.00 g Cu

10.00 g/L, for 1L standard solution, analytical standard

Synonym(s):

Cupric chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Cu
CAS Number:
Molecular Weight:
134.45
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

packaging

ampule of

concentration

10.00 g/L

solubility

H2O: soluble

SMILES string

Cl[Cu]Cl

InChI

1S/2ClH.Cu/h2*1H;/q;;+2/p-2

Inchi Key

ORTQZVOHEJQUHG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Preparation Note

prepared with CuCl2.2H2O, HCl and H2O

Pictograms

CorrosionEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William T Triplett et al.
Magnetic resonance in medicine, 72(1), 8-19 (2013-09-06)
The relationship between fat fractions (FFs) determined based on multiple TE, unipolar gradient echo images and (1) H magnetic resonance spectroscopy (MRS) was evaluated using different models for fat-water decomposition, signal-to-noise ratios, and excitation flip angles. A combination of single-voxel
Kenji Funaki et al.
Organic letters, 14(24), 6186-6189 (2012-12-12)
Direct arylation of thiophenes and benzothiophenes with aryltrimethylsilanes was effectively catalyzed by PdCl(2)(MeCN)(2) in the presence of CuCl(2) as an oxidant. The reaction preferentially occurred at the β-position of both thiophenes and benzothiophenes.
Su-Jeong Kim et al.
Free radical biology & medicine, 53(4), 936-950 (2012-06-12)
Loss-of-function mutations in the PARK7/DJ-1 gene cause early onset autosomal-recessive Parkinson disease. DJ-1 has been implicated in protection of neurons from oxidative stress and in regulation of transcriptional activity. However, whether there is a relationship between the subcellular localization of
Mónica João Barros Amorim et al.
Environmental pollution (Barking, Essex : 1987), 164, 164-168 (2012-03-01)
Environmental effects of copper nanoparticles are little studied in terrestrial ecosystems. In the present article, the toxicity of copper nanoparticles (Cu-NP) on the enchytraeid Enchytraeus albidus is compared to the toxicity of a copper-salt (CuCl(2)). The effect parameters studied were
Landon John G Edgar et al.
Organic letters, 14(16), 4226-4229 (2012-08-01)
Glycosyl 1-phosphates enriched in the α-anomer are obtained without the use of protecting groups in two steps starting from the free hemiacetal. Condensation of free hemiacetals with toluenesulfonylhydrazide yields a range of glycosylsulfonohydrazide donors which can be oxidized using cupric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service