03368
Nickel atomic spectroscopy standard concentrate 10.00 g Ni
10.00 g/L, for 1L standard solution, analytical standard
Synonym(s):
Nickel(II) sulfate solution
About This Item
grade
analytical standard
shelf life
limited shelf life, expiry date on the label
packaging
ampule of
concentration
10.00 g/L
solubility
H2O: soluble
SMILES string
[Ni]
InChI
1S/Ni
InChI key
PXHVJJICTQNCMI-UHFFFAOYSA-N
Preparation Note
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1
target_organs
respiratory tract irritation
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service