03030
2-Pentylfuran
analytical standard
Synonym(s):
2-Amylfuran
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥97.0% (GC)
shelf life
limited shelf life, expiry date on the label
refractive index
n20/D 1.448 (lit.)
n20/D 1.448
bp
64-66 °C/23 mmHg (lit.)
density
0.883 g/mL at 20 °C (lit.)
0.886 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
neat
SMILES string
CCCCCc1ccco1
InChI
1S/C9H14O/c1-2-3-4-6-9-7-5-8-10-9/h5,7-8H,2-4,6H2,1H3
InChI key
YVBAUDVGOFCUSG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 3
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
114.8 °F
flash_point_c
46 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service