Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

02946

Sigma-Aldrich

Ethylamine

anhydrous, ≥99.5%

Synonym(s):

Aminoethane, Monoethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5NH2
CAS Number:
Molecular Weight:
45.08
Beilstein/REAXYS Number:
505933
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:

vapor density

1.56 (15 °C, vs air)

vapor pressure

874 mmHg ( 20 °C)

assay

≥99.5%

autoignition temp.

721 °F

quality

anhydrous

expl. lim.

14 %
3.5-14 %

impurities

≤0.4% water

bp

16.6 °C (lit.)

mp

-81 °C (lit.)

density

0.680 g/mL at 20 °C (lit.)
0.689 g/mL at 25 °C (lit.)

SMILES string

CCN

InChI

1S/C2H7N/c1-2-3/h2-3H2,1H3

InChI key

QUSNBJAOOMFDIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Cylinder with net ~15 kg

Other Notes

Sales restrictions may apply

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Gas 1 - Press. Gas Liquefied gas - STOT SE 3

target_organs

Respiratory system

Storage Class

2A - Gases

wgk_germany

WGK 1

flash_point_f

-34.6 °F - closed cup

flash_point_c

-37 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Stacey F Bent et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(3), 956-960 (2010-11-12)
Surface functionalization of semiconductors has been the backbone of the newest developments in microelectronics, energy conversion, sensing device design, and many other fields of science and technology. Over a decade ago, the notion of viewing the surface itself as a
Paige E Mahaney et al.
Journal of medicinal chemistry, 51(13), 4038-4049 (2008-06-19)
Further exploration of the cycloalkanol ethylamine scaffold, of which venlafaxine ( 1) is a member, was undertaken to develop novel and selective norepinephrine reuptake inhibitors (NRIs) for evaluation in a variety of predictive animal models. These efforts led to the
Yoko Okubo et al.
Journal of bacteriology, 192(7), 1813-1823 (2010-02-02)
Methylobacterium extorquens AM1 is a facultative methylotroph capable of growth on both single-carbon and multicarbon compounds. Mutants defective in a pathway involved in converting acetyl-coenzyme A (CoA) to glyoxylate (the ethylmalonyl-CoA pathway) are unable to grow on both C(1) and
Riccardo Di Corato et al.
ACS nano, 5(2), 1109-1121 (2011-01-12)
Trifunctional polymer nanobeads are prepared by destabilization of a mixture of magnetic nanoparticles, quantum dots, and an amphiphilic polymer, followed by functionalization of the bead surface with folic acid molecules. The distribution of the nanoparticles within the nanobeads can be
Veronica Sandgren et al.
Bioorganic & medicinal chemistry, 20(14), 4377-4389 (2012-06-16)
A series of P1-P3 linked macrocyclic BACE-1 inhibitors containing a hydroxyethylamine (HEA) isostere scaffold has been synthesized. All inhibitors comprise a toluene or N-phenylmethanesulfonamide P2 moiety. Excellent BACE-1 potencies, both in enzymatic and cell-based assays, were observed in this series

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service