Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

02673

Supelco

Chromium(VI) atomic spectroscopy standard concentrate 1.00 g Cr(VI)

1.00 g/L, for 1L standard solution, analytical standard

Synonym(s):

Potassium chromate solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CrO4
CAS Number:
Molecular Weight:
194.19
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

packaging

ampule of

solubility

H2O: soluble

SMILES string

[K+].[K+].[O-][Cr]([O-])(=O)=O

InChI

1S/Cr.2K.4O/q;2*+1;;;2*-1

InChI key

XMXNVYPJWBTAHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

prepared with K2CrO4 and H2O

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B Inhalation - Muta. 1B - Skin Sens. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subhendra N Mattagajasingh et al.
Journal of applied toxicology : JAT, 28(8), 987-997 (2008-07-11)
Hexavalent chromium [Cr(VI)] compounds are Group-I human carcinogens. Cr(VI)-induced DNA-protein crosslinks (DPCs) have been implicated in the mutagenic and carcinogenic effects of Cr(VI). Although multiple mechanisms have been suggested for Cr(VI)-induced DNA-protein crosslinking, the mechanism of formation of DNA-protein crosslinks
Xue Zhou et al.
Toxicology and applied pharmacology, 236(1), 78-84 (2009-04-18)
Occupational exposure to nickel (Ni), chromium (Cr), and arsenic (As) containing compounds has been associated with lung cancer and other adverse health effects. Their carcinogenic properties may be attributable in part, to activation and/or repression of gene expression induced by
Cecylia Wardak
Journal of hazardous materials, 186(2-3), 1131-1135 (2010-12-21)
A new polyvinylchloride membrane sensor for Pb(2+) with solid contact based on ionic liquid has been prepared. The electrode shows a Nernstian response for lead ions over a wide concentration range (1×10(-8) to 1×10(-1) mol L(-1)) and the slope of
Saturnino Spiga et al.
Brain structure & function, 216(3), 171-182 (2011-04-05)
Visualization of neuronal elements is of fundamental importance in modern neuroscience. Golgi-Cox impregnation is a widely employed method that provides detailed information about morphological characteristics of neurons, but none regarding their neurochemical features. Immunocytochemical procedures, on the other hand, can
Nejla Soudani et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 63(6), 541-548 (2010-05-25)
Chromium, a major environmental pollutant, is known for its wide toxic manifestations. The present experiment pertains to the protective role of selenium (Se) against K(2)Cr(2)O(7)-induced hepatotoxicity. Female Wistar rats were divided into four groups of six each: group I served

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service