Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

02653

Supelco

Molybdenum atomic spectroscopy standard concentrate 1.00 g Mo

1.00 g/L, for 1L standard solution, analytical standard

Synonym(s):

Ammonium molybdate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

packaging

ampule of

solubility

H2O: soluble

SMILES string

[Mo]

InChI

1S/Mo

InChI key

ZOKXTWBITQBERF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

glass ampoule

Preparation Note

prepared with (NH4)6Mo7O24 and H2O

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[The hygienic regulation of harmful substances in the atmosphere taking into account body adaptational reactions].
V M Pazynich et al.
Gigiena i sanitariia, (1)(1), 11-14 (1993-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service