Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

02588

Supelco

Nickel atomic spectroscopy standard concentrate 1.00 g Ni

1.00 g/L, for 1L standard solution, analytical standard

Synonym(s):

Nickel(II) chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

reaction suitability

reagent type: catalyst
core: nickel

packaging

ampule of

solubility

H2O: soluble

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

prepared with NiCl2.6H2O and H2O

pictograms

Health hazardEnvironment

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 - STOT RE 2 Inhalation

target_organs

Lungs

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingwen Yu et al.
Bio-medical materials and engineering, 22(1-3), 151-157 (2012-07-07)
Dental cast alloys have been widely used in modern dental restoration. However, due to oral complex environment and individual differences, continuous corrosion of alloys occurs. As a result, metal ions like nickel ion (Ni3+) can be released from alloys for
Terran E Bergdale et al.
Journal of biotechnology, 161(3), 181-189 (2012-07-14)
Microbiologically induced calcium carbonate precipitation (MICCP) is a naturally occurring biological process that has shown its potential in remediation of a wide range of structural damages including concrete cracks. In this study, genetically engineered microorganisms, capable of producing extracellular polymeric
Rebekka Kubisch et al.
Sensors (Basel, Switzerland), 12(3), 3370-3393 (2012-06-28)
Pollution of drinking water sources represents a continuously emerging problem in global environmental protection. Novel techniques for real-time monitoring of water quality, capable of the detection of unanticipated toxic and bioactive substances, are urgently needed. In this study, the applicability
Steven J Ryan et al.
PloS one, 7(4), e35320-e35320 (2012-05-09)
The basolateral complex of the amygdala (BLA) is a critical component of the neural circuit regulating fear learning. During fear learning and recall, the amygdala and other brain regions, including the hippocampus and prefrontal cortex, exhibit phase-locked oscillations in the
Abhay K Pandey et al.
Neuroscience letters, 517(2), 128-132 (2012-05-09)
Bisphenol-A (BPA), a toxic chemical from polycarbonate plastics, is known for behavioural and neural abnormalities. These neuro-behavioural changes reflect the changes in neural activity. However the effect of BPA on nerve action potential is not available. Therefore, present investigation was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service