Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

02380

Sigma-Aldrich

1,2-Ethanedithiobis(trimethylsilane)

≥97.0%

Synonym(s):

1,2-Bis(trimethylsilylthio)ethane, Ethylenedithiobis(trimethylsilane), S,S′-Bis(trimethylsilyl)-1,2-ethanedithiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H22S2Si2
CAS Number:
Molecular Weight:
238.56
Beilstein/REAXYS Number:
2037789
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0%

refractive index

n20/D 1.495

bp

93-94 °C/0.5 mmHg (lit.)

density

0.928 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(C)SCCS[Si](C)(C)C

InChI

1S/C8H22S2Si2/c1-11(2,3)9-7-8-10-12(4,5)6/h7-8H2,1-6H3

InChI key

CBRFJNLREFDKBD-UHFFFAOYSA-N

Other Notes

Reagent used for the kinetic, regioselective thioacetalization of carbonyl compounds[1][2]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

152.6 °F - closed cup

flash_point_c

67 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.P. Tanis et al.
Tetrahedron Letters, 28, 2495-2495 (1987)
D.A. Evans et al.
Journal of the American Chemical Society, 99, 5009-5009 (1977)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service