02359
Poly(methacrylic acid sodium salt)
analytical standard, 75,100
Synonym(s):
Poly(sodium methacrylate)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
grade
analytical standard
Quality Level
shelf life
limited shelf life, expiry date on the label
mol wt
Mn ~73300
Mp ~78300
Mw ~75100
technique(s)
gel permeation chromatography (GPC): suitable
Mw/Mn
~1.02
storage temp.
2-8°C
InChI
1S/C4H6O2.Na/c1-3(2)4(5)6;/h1H2,2H3,(H,5,6);/q;+1/p-1
InChI key
SONHXMAHPHADTF-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service