Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

02356

Supelco

Poly(methacrylic acid sodium salt)

analytical standard, 18,500

Synonym(s):

Poly(sodium methacrylate)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

mol wt

Mp ~18500
Mw ~18600

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

<1.20

storage temp.

2-8°C

InChI

1S/C4H6O2.Na/c1-3(2)4(5)6;/h1H2,2H3,(H,5,6);/q;+1/p-1

InChI key

SONHXMAHPHADTF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Poly(methacrylic acid sodium salt) is used as an analytical standard in the development of nebulized amphotericin B-polymethacrylic acid nanoparticles for use as pre-exposure prophylaxis agents to deliver anti-microbial drugs to the lung and inhibition of fungal infections caused by Aspergillus species.[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nebulised amphotericin B-polymethacrylic acid nanoparticle prophylaxis prevents invasive aspergillosis
Shirkhani K, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 11(5), 1217-1226 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service