Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

02134

Supelco

6-O-Palmitoyl-L-ascorbic acid

analytical standard

Synonym(s):

L-Ascorbyl palmitate, Ascorbic acid 6-palmitate, E304, L-Ascorbic acid 6-hexadecanoate, NSC 402451

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H38O7
CAS Number:
Molecular Weight:
414.53
Beilstein/REAXYS Number:
96552
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

Quality Level

grade

analytical standard

assay

≥98.5% (HPLC)

form

powder or crystals

shelf life

limited shelf life, expiry date on the label

loss

≤2.0% loss on drying

color

white to light beige

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

InChI

1S/C22H38O7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(24)28-16-17(23)21-19(25)20(26)22(27)29-21/h17,21,23,25-26H,2-16H2,1H3/t17-,21+/m0/s1

InChI key

QAQJMLQRFWZOBN-LAUBAEHRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alex C M Oliveira et al.
Nutrients, 6(5), 2059-2076 (2014-05-27)
The beneficial health effects of a diet rich in n-3 long chain polyunsaturated fatty acids (n-3 LC-PUFA) have been extensively researched in recent years. Marine oils are an important dietary source of n-3 LC-PUFA, being especially rich in two of
Ascorbyl palmitate and its synergism to tocopherols.
Karl-Heinz Wagner et al.
Forum of nutrition, 56, 347-348 (2005-04-06)
Li Xiao et al.
Molecular and cellular biochemistry, 394(1-2), 247-259 (2014-06-06)
The aim of this study was to investigate protective effects of the lipophilic vitamin C derivative, 6-o-palmitoylascorbate (PlmtVC), against X-ray radiation-induced damages including cell death, DNA double-strand breaks (DSBs), lipid peroxidation, and protein carbonylation in human lymphocytes HEV0082, and the
Mostafa Nakach et al.
International journal of pharmaceutics, 476(1-2), 277-288 (2014-10-04)
This paper describes a systematic approach to select optimum stabilizer for the preparation of nano-crystalline suspensions of an active pharmaceutical ingredient (API). The stabilizer can be either a dispersant or a combination of dispersant and wetting agent. The proposed screening
Lingyan Kong et al.
Carbohydrate polymers, 111, 256-263 (2014-07-20)
In the present study, we introduce a simple method to prepare inclusion complexes by "inserting" guest molecules into preformed "empty" V-type amylose helices. Ascorbyl palmitate (AscP) was used as a model guest material to investigate the effect of solvent environment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service