02116
Copper(I) bromide
≥97.0%
Synonym(s):
Cuprous bromide
About This Item
Recommended Products
grade
for analytical purposes
assay
≥97.0%
form
powder
reaction suitability
reagent type: catalyst
core: copper
impurities
≤0.02% insoluble in HCl
≤0.3% subst. not precipitated by H2S (as SO4)
≤4.5% CuBr2
mp
504 °C (lit.)
density
4.71 g/mL at 25 °C (lit.)
anion traces
sulfate (SO42-): ≤5000 mg/kg
cation traces
As: ≤50 mg/kg
Fe: ≤500 mg/kg
SMILES string
[Cu]Br
InChI
1S/BrH.Cu/h1H;/q;+1/p-1
InChI key
NKNDPYCGAZPOFS-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service