Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01972

Supelco

Serdolit CG-400 I

Cl--Form, strongly basic, 100-200 mesh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
SB.52

form

beads

quality

strongly basic

technique(s)

LPLC: suitable

loss

≤18% loss on drying

matrix

polystyrene-divinylbenzene

matrix active group

quaternary ammonium

particle size

100-200 mesh

capacity

1.9 meq/mL

separation technique

anion exchange

Legal Information

Serdolit is a trademark of Serva Electrophoresis GmbH

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service