Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

01961

Sigma-Aldrich

1,5,7-Triazabicyclo[4.4.0]dec-5-ene bound to polystyrene

extent of labeling: ~3 mmol/g loading

Synonym(s):

1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine bound to polystyrene, 7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene on polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: proton reactive

extent of labeling

~3 mmol/g loading

matrix

crosslinked with 2% DVB

particle size

16-50 mesh

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service