01428
Stable Isotope Labeled Amino Acid Mix Solution 2
500-2500 nmol/mL in 0.1 M HCl, certified reference material, TraceCERT®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
product line
TraceCERT®
form
liquid
shelf life
limited shelf life, expiry date on the label
concentration
500-2500 nmol/mL in 0.1 M HCl
storage temp.
−20°C
General description
11 amino acids in 0.1M hydrochloric acid
0.5 mM each: L-Alanine-13C3,15N, L-Arginine-13C6, L-Aspartic acid-13C4, L-Glutamic acid-13C5, L-Leucine-13C6,15N, L-Methionine-13C5,15N, L-Phenyl-13C6-alanine, L-Proline-13C5, L-Tyrosine-(phenyl-13C6), L-Valine-13C5
and 2.5mM Glycine-13C2,15N
0.5 mM each: L-Alanine-13C3,15N, L-Arginine-13C6, L-Aspartic acid-13C4, L-Glutamic acid-13C5, L-Leucine-13C6,15N, L-Methionine-13C5,15N, L-Phenyl-13C6-alanine, L-Proline-13C5, L-Tyrosine-(phenyl-13C6), L-Valine-13C5
and 2.5mM Glycine-13C2,15N
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Legal Information
TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
signalword
Warning
hcodes
pcodes
Hazard Classifications
Met. Corr. 1
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service