Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01375

Sigma-Aldrich

Newman′s Stain Solution, modified

Synonym(s):

Newman-Lampert Stain Solution, modified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
MA.02

product line

BioChemika

Quality Level

form

liquid

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | staining: suitable

suitability

suitable for microscopy
bacteria

application(s)

food and beverages
hematology
histology

storage temp.

room temp

General description

Newman`s stain modified is designed to find somatic cell count in milk or certain milk products[1]

Components

Composition:
Methylene blue chloride 0.6 g, Ethanol (95%) 52 mL, Tetrachloroethane 44 mL, Glacial acetic acid 4 ml

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

68.0 °F - closed cup

flash_point_c

20 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.J. Paape et al.
Journal of Dairy Science, 84E, 237-237 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service