Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01290190

Geranyl acetate

primary reference standard

Synonym(s):

trans-3,7-Dimethyl-2,6-octadien-1-yl acetate, trans-3,7-Dimethyl-2,6-octadienyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
196.29
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

vapor density

6.8 (vs air)

vapor pressure

0.07 mmHg ( 20 °C)

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

refractive index

n20/D 1.461 (lit.)

bp

138 °C/25 mmHg (lit.)

storage temp.

−20°C

SMILES string

CC(=O)OC\C=C(/C)CC\C=C(/C)C

InChI

1S/C12H20O2/c1-10(2)6-5-7-11(3)8-9-14-12(4)13/h6,8H,5,7,9H2,1-4H3/b11-8+

InChI key

HIGQPQRQIQDZMP-DHZHZOJOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

219.2 °F - closed cup

flash_point_c

104 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service