Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01192

Sigma-Aldrich

Magnesium fluoride

Synonym(s):

Sellaite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgF2
CAS Number:
Molecular Weight:
62.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

powder

density

3.15 g/mL at 25 °C (lit.)

SMILES string

F[Mg]F

InChI

1S/2FH.Mg/h2*1H;/q;;+2/p-2

InChI key

ORUIBWPALBXDOA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takehiro Shinoda et al.
Nature, 459(7245), 446-450 (2009-05-22)
Sodium-potassium ATPase is an ATP-powered ion pump that establishes concentration gradients for Na(+) and K(+) ions across the plasma membrane in all animal cells by pumping Na(+) from the cytoplasm and K(+) from the extracellular medium. Such gradients are used
Ji-Hoon Jo et al.
Journal of materials science. Materials in medicine, 22(11), 2437-2447 (2011-09-13)
Hydroxyapatite (HA) was coated onto pure magnesium (Mg) with an MgF(2) interlayer in order to reduce the surface corrosion rate and enhance the biocompatibility. Both MgF(2) and HA were successfully coated in sequence with good adhesion properties using the fluoride
Naoto Shibata et al.
The journal of physical chemistry. B, 115(35), 10553-10559 (2011-08-19)
A series of magnesium fluorides (MgF(n)(2-n)), multiply charged anions, in the gas phase and in aqueous solution were theoretically studied with a hybrid approach of quantum chemistry and statistical mechanics, called RISM-SCF-SEDD theory. In the gas phase, MgF(3)(-) is the
Enrique Marcos et al.
Proteins, 78(11), 2405-2411 (2010-07-06)
Enzymes catalyzing phosphoryl transfer reactions are extremely efficient and are involved in crucial biochemical processes. The mechanisms of these enzymes are complex due to the diversity of substrates that are involved. The reaction can proceed through a pentacoordinated phosphorus species
Manwen Yao et al.
Acta biomaterialia, 4(6), 2016-2027 (2008-07-29)
The effects of intermediate dielectric films on multilayer surface plasmon resonance (SPR) behavior were studied in terms of biosensing applications. Ten simple and complex oxides and fluoride, including MgF(2) and MgO, SiO(2), TiO(2) and complex PZT family dielectric materials, were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service