Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01110

Sigma-Aldrich

Ammonium hydrogen difluoride

puriss., ≥97.5%

Synonym(s):

Ammonium bifluoride, Etching powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4HF2
CAS Number:
Molecular Weight:
57.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

puriss.

assay

≥97.5%

impurities

≤0.001% heavy metals (as Pb)
≤0.03% hexafluorosilicate (as (NH4)2SiF6)

mp

125 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg
sulfite (SO32-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Fe: ≤100 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg

SMILES string

N.F.F

InChI

1S/2FH.H3N/h2*1H;1H3

InChI key

KVBCYCWRDBDGBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesCorrosion

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroko Sano et al.
PLoS genetics, 11(5), e1005209-e1005209 (2015-05-29)
The coordination of growth with nutritional status is essential for proper development and physiology. Nutritional information is mostly perceived by peripheral organs before being relayed to the brain, which modulates physiological responses. Hormonal signaling ensures this organ-to-organ communication, and the
Sebastian Ribi et al.
Oncotarget, 6(10), 7727-7740 (2015-03-13)
Somatic mutations of TP53 are among the most common in cancer and germline mutations of TP53 (usually missense) can cause Li-Fraumeni syndrome (LFS). Recently, recurrent genomic rearrangements in intron 1 of TP53 have been described in osteosarcoma (OS), a highly
Jinhong Li et al.
The New phytologist, 208(1), 137-148 (2015-04-14)
Heteromorphic flower development in Primula is controlled by the S locus. The S locus genes, which control anther position, pistil length and pollen size in pin and thrum flowers, have not yet been characterized. We have integrated S-linked genes, marker
Nick Orr et al.
Human molecular genetics, 24(10), 2966-2984 (2015-02-06)
We recently identified a novel susceptibility variant, rs865686, for estrogen-receptor positive breast cancer at 9q31.2. Here, we report a fine-mapping analysis of the 9q31.2 susceptibility locus using 43 160 cases and 42 600 controls of European ancestry ascertained from 52
Wesley C Warren et al.
Genome research, 25(12), 1921-1933 (2015-09-18)
We describe a genome reference of the African green monkey or vervet (Chlorocebus aethiops). This member of the Old World monkey (OWM) superfamily is uniquely valuable for genetic investigations of simian immunodeficiency virus (SIV), for which it is the most

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service