Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

00873

Sigma-Aldrich

(R)-α-Acetoxyphenylacetonitrile

≥97.0% (sum of enantiomers, GC)

Synonym(s):

(R)-α-Cyanobenzyl acetate, O-Acetyl-D-mandelonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
Beilstein/REAXYS Number:
4293296
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0% (sum of enantiomers, GC)

optical activity

[α]20/D +8.0±0.5°, c = 10% in chloroform

refractive index

n20/D 1.506

density

1.115 g/mL at 20 °C (lit.)

SMILES string

CC(=O)O[C@@H](C#N)c1ccccc1

InChI

1S/C10H9NO2/c1-8(12)13-10(7-11)9-5-3-2-4-6-9/h2-6,10H,1H3/t10-/m0/s1

InChI key

MUXDFYRMYMEGCM-JTQLQIEISA-N

Application

(R)-α-Acetoxyphenylacetonitrile can be used as a model compound in the iodine promoted as well as t-BuONO-assisted secondary amides synthesis via an aryl N-addition reaction to the -CN group of nitriles.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iodine-mediated aryl transfer reaction from arylhydrazine hydrochlorides to nitriles
Zhang Z, et al.
Tetrahedron, 75(26), 3628-3635 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service