Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00846

Sigma-Aldrich

(1R,2R)-trans-2-Acetoxy-1-cyclopentanol

≥99.0% (sum of enantiomers, GC)

Synonym(s):

(1R)-trans-1,2-Cyclopentanediol monoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O3
CAS Number:
Molecular Weight:
144.17
Beilstein/REAXYS Number:
5730491
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

assay

≥99.0% (sum of enantiomers, GC)

optical activity

[α]20/D +31±2°, c = 1% in chloroform

optical purity

enantiomeric ratio: ≥98:2 (GC)

refractive index

n20/D 1.455

density

1.102 g/mL at 20 °C (lit.)

SMILES string

CC(=O)O[C@@H]1CCC[C@H]1O

InChI

1S/C7H12O3/c1-5(8)10-7-4-2-3-6(7)9/h6-7,9H,2-4H2,1H3/t6-,7-/m1/s1

InChI key

YRJFZPSFQDTFRK-RNFRBKRXSA-N

Application

Chiral alcohol derived from (R,R)-trans-1,2-cyclopentanediol

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service