Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00787

Sigma-Aldrich

N-Benzyl-N′-cyclohexylcarbodiimide, polymer-bound

100-300 mesh, extent of labeling: ~1.3 mmol/g N loading

Synonym(s):

N,N′-Dicyclohexylcarbodiimide, polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: carboxyl reactive

extent of labeling

~1.3 mmol/g N loading

matrix

cross-linked with 2% DVB

particle size

100-300 mesh

Looking for similar products? Visit Product Comparison Guide

Other Notes

Immobilized carbodiimide for peptide coupling allowing simple removal of excess reagent and the urea by-product.[1]

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N.M. Weinshenker et al.
Organic Syntheses, VI, 951-951 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service