Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00721

Supelco

4-(Dimethyl-d6-amino)benzoyl chloride

for HPLC derivatization, ≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D6H4ClNO
CAS Number:
Molecular Weight:
189.67
MDL number:
UNSPSC Code:
41121800
PubChem Substance ID:
NACRES:
NB.21

grade

derivatization grade ((HPLC))
for HPLC derivatization

Quality Level

assay

≥97.0% (HPLC)

mp

150-153 °C

suitability

complies for LC-MS

storage temp.

2-8°C

SMILES string

ClC(C1=CC=C(N(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C1)=O

InChI

1S/C9H10ClNO/c1-11(2)8-5-3-7(4-6-8)9(10)12/h3-6H,1-2H3/i1D3,2D3

InChI key

UGJDXRVQCYBXAJ-WFGJKAKNSA-N

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshio Suzuki et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 20(3), 475-482 (2004-04-08)
Novel labeling reagents, called MS probes, which possess a positively charged quaternary amine moiety and can transform a neutral analyte into a charged compound by simply mixing with the analyte and allowing the mixture to stand from several minutes to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service