Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00609

Supelco

Octocrylene-(2-ethyl-d5-hexyl-2,3,3,4,4,5,5,6,6,6-d10)

Octocrylene-(2-ethyl-d5-hexyl-2,3,3,4,4,5,5,6,6,6-d10)

analytical standard

Synonym(s):

Octocrylene-d15, [2-(Ethyl-d5)-hexyl-2,3,3,4,4,5,5,6,6,6-d10] 2-cyano-3,3-diphenylacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24D15H12NO2
Molecular Weight:
376.57
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

isotopic purity

≥97 atom % D

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

Active Filters

  1. South
    • Review 1
    • Votes 0
    1 out of 5 stars.

    Useless ISTD, glad it's discontinued

    The compound fragments into the same daughter ions as octocrylene. The deuteriums are on the wrong side of the molecule, so why 15 were added I have no idea.

    Helpful?

    1. Response from MilliporeSigma:

      Thank you for your review. We are sorry to hear that your experience did not match your product expectations. We encourage customers who experience problems with our products to contact us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service