Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00589

Sigma-Aldrich

N-Trityl-1,2-ethanediamine hydrobromide

>97.0% (AT)

Synonym(s):

N-Trityl-1,2-diaminoethane hydrobromide, N-Tritylethylenediamine hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N2 · HBr
CAS Number:
Molecular Weight:
383.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

>97.0% (AT)

reaction suitability

reagent type: cross-linking reagent

impurities

≤3.0% water

mp

187-189 °C

storage temp.

2-8°C

SMILES string

Br[H].NCCNC(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C21H22N2.BrH/c22-16-17-23-21(18-10-4-1-5-11-18,19-12-6-2-7-13-19)20-14-8-3-9-15-20;/h1-15,23H,16-17,22H2;1H

InChI key

ILSWDBONXPWRBT-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Pavić et al.
European journal of medicinal chemistry, 86, 502-514 (2014-09-10)
Novel primaquine semicarbazides 7a-l and ureas 9a-g with modified benzhydryl, trityl, phenyl or hydroxyalkyl substituents were prepared and evaluated for cytostatic and antioxidative activities. Two synthetic approaches for preparation of the title semicarbazides were applied, both having certain advantages. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service