About This Item
Recommended Products
assay
≥99.998%
SMILES string
O=O
InChI
1S/O2/c1-2
InChI key
MYMOFIZGZYHOMD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
filling pressure at 15°C: 11 bar; content (15°C, 1 bar): 11 l
Other Notes
signalword
Danger
hcodes
Hazard Classifications
Ox. Gas 1 - Press. Gas Compr. Gas
Storage Class
2A - Gases
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service