Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00474

Sigma-Aldrich

Nitrogen

≥99.999%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
N2
CAS Number:
Molecular Weight:
28.01
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
32140601
PubChem Substance ID:
NACRES:
NA.23

vapor density

0.97 (vs air)

assay

≥99.999%

bp

−196 °C (lit.)

mp

−210 °C (lit.)

SMILES string

N#N

InChI

1S/N2/c1-2

InChI key

IJGRMHOSHXDMSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

pressure tin
filling pressure at 15°C: 11 bar; content (15°C, 1 bar): 11 l

Other Notes

Sales restrictions may apply

pictograms

Gas cylinder

signalword

Warning

hcodes

Hazard Classifications

Press. Gas Compr. Gas

Storage Class

2A - Gases

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piero Amodio et al.
Hepatology (Baltimore, Md.), 58(1), 325-336 (2013-03-09)
Nitrogen metabolism plays a major role in the development of hepatic encephalopathy (HE) in patients with cirrhosis. Modulation of this relationship is key to the management of HE, but is not the only nutritional issue that needs to be addressed.
Florence Guida et al.
Human molecular genetics, 24(8), 2349-2359 (2015-01-04)
Several studies have recently identified strong epigenetic signals related to tobacco smoking. However, an aspect that did not receive much attention is the evolution of epigenetic changes with time since smoking cessation. We conducted a series of epigenome-wide association studies
Wally C van Heeswijk et al.
Microbiology and molecular biology reviews : MMBR, 77(4), 628-695 (2013-12-04)
We present a comprehensive overview of the hierarchical network of intracellular processes revolving around central nitrogen metabolism in Escherichia coli. The hierarchy intertwines transport, metabolism, signaling leading to posttranslational modification, and transcription. The protein components of the network include an
T Jickells et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 368(1621), 20130115-20130115 (2013-05-29)
Atmospheric organic nitrogen (ON) appears to be a ubiquitous but poorly understood component of the atmospheric nitrogen deposition flux. Here, we focus on the ON components that dominate deposition and do not consider reactive atmospheric gases containing ON such as
Long-Feng Xu et al.
PloS one, 9(8), e103698-e103698 (2014-08-02)
In South China (Gejiu City, Yunnan Province), lung cancer incidence and associated mortality rate is the most prevalent and observed forms of cancer. Lung cancer in this area is called Gejiu squamous cell lung carcinoma (GSQCLC). Research has demonstrated that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service