About This Item
Recommended Products
assay
≥99.995%
SMILES string
[Xe]
InChI
1S/Xe
InChI key
FHNFHKCVQCLJFQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
filling pressure at 15°C: 11 bar; content (15°C, 1 bar): 11 L
Other Notes
signalword
Warning
hcodes
pcodes
Hazard Classifications
Press. Gas Liquefied gas
Storage Class
2A - Gases
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service