Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00437

Supelco

2,4-Di-tert-butylphenol

reference material

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H3OH
CAS Number:
Molecular Weight:
206.32
Beilstein/REAXYS Number:
1910383
MDL number:
UNSPSC Code:
41116107

grade

reference material

vapor pressure

1 mmHg ( 84.5 °C)

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

bp

265 °C (lit.)

mp

53-56 °C (lit.)

SMILES string

CC(C)(C)c1ccc(O)c(c1)C(C)(C)C

InChI

1S/C14H22O/c1-13(2,3)10-7-8-12(15)11(9-10)14(4,5)6/h7-9,15H,1-6H3

InChI key

ICKWICRCANNIBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

pictograms

CorrosionEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

264.2 °F

flash_point_c

129 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service