Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00346

Sigma-Aldrich

Fmoc-β-(3-thienyl)-Ala-OH

≥98.0% (HPLC)

Synonym(s):

(S)-2-(Fmoc-amino)-3-(3-thienyl)propionic acid, Fmoc-3-(3-thienyl)-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19NO4S
CAS Number:
Molecular Weight:
393.46
Beilstein/REAXYS Number:
9512976
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Quality Level

assay

≥98.0% (HPLC)

form

powder with small lumps

color

white

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](Cc1ccsc1)NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C22H19NO4S/c24-21(25)20(11-14-9-10-28-13-14)23-22(26)27-12-19-17-7-3-1-5-15(17)16-6-2-4-8-18(16)19/h1-10,13,19-20H,11-12H2,(H,23,26)(H,24,25)/t20-/m0/s1

InChI key

LSBZJMRHROCYGY-FQEVSTJZSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service