Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00290595

(−)-Carvone

primary reference standard

Synonym(s):

(−)-p-Mentha 6,8-diene 2-one, (R)-5-Isopropenyl-2-methyl-2-cyclohexenone, Carvol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Beilstein/REAXYS Number:
2206714
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

bp

228-230 °C (lit.)

density

0.960 g/mL at 20 °C (lit.)

SMILES string

CC(=C)[C@@H]1CC=C(C)C(=O)C1

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)10(11)6-9/h4,9H,1,5-6H2,2-3H3/t9-/m1/s1

InChI key

ULDHMXUKGWMISQ-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

192.2 °F - closed cup

flash_point_c

89 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hisahiro Hagiwara
Natural product communications, 8(7), 935-948 (2013-08-29)
This review article is focused on the total syntheses of heterocyclic sesquiterpenoids starting from carvone since 1996.
Qin Kang et al.
Biochemical and biophysical research communications, 459(3), 498-503 (2015-03-10)
Transient receptor potential (TRP) channels in the spinal dorsal horn lamina II (substantia gelatinosa; SG), which are involved in the modulation of nociceptive transmission, have not yet been fully examined in property. Activation of the TRP channels by various plant-derived
Lee Martin et al.
Dalton transactions (Cambridge, England : 2003), 44(13), 6219-6223 (2015-03-05)
The first radical-cation salts in the extensive family (BEDT-TTF)x[(A)M(C2O4)3]·Guest containing lithium as the counter cation have been synthesized and characterised.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service