Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00234-1KG

Amberlite IRN150 hydrogen and hydroxide form

hydrogen and hydroxide form

Sign Into View Organizational & Contract Pricing

Select a Size

00234-1KG
$311.00

$311.00


Available to ship onMay 01, 2025Details



Select a Size

Change View
00234-1KG
$311.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817

$311.00


Available to ship onMay 01, 2025Details


description

cross-linkage 8%
strongly basic

form

beads

parameter

60 °C max. temp.

moisture

49-55% (cation)
54-60% (anion)

loss

~55% loss on drying, 110 °C

matrix

styrene/divinylbenzene (gel)

matrix active group

sulfonic acid functional group, trimethylammonium

particle size

580-680 μm (anion exchanger)
600-700 μm (cation exchanger)

operating pH

0-14

capacity

1.2 meq/g by dry weight(anion)
1.9 meq/g by dry weight(cation)

Looking for similar products? Visit Product Comparison Guide

General description

Mixed bed resin for primary water chemistry control in once-through systems. Useful in industrial water treatment where as-supplied resin must have absolute minimum of ionic and non-ionic contamination.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service