Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40-4004

Sigma-Aldrich

Methyl Orange-Xylene cyanole FF phenolphthalein solution

pH 4.2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710
Pricing and availability is not currently available.

form

liquid

availability

available only in Japan

dilution

(for analytical testing)

pH

4.2

Suitability

for titration

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

55.4 °F

flash_point_c

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service