Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40-2015

Sigma-Aldrich

Bromocresol Green solution

pH 3.8-5.4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14Br4O5S
CAS Number:
Molecular Weight:
698.01
Beilstein/REAXYS Number:
372527
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

form

liquid

availability

available only in Japan

pH

3.8-5.4

SMILES string

O=S1(C2=CC=CC=C2C(C3=CC(Br)=C(O)C(Br)=C3C)(C4=CC(Br)=C(O)C(Br)=C4C)O1)=O

InChI

1S/C21H14Br4O5S/c1-9-12(7-14(22)19(26)17(9)24)21(13-8-15(23)20(27)18(25)10(13)2)11-5-3-4-6-16(11)31(28,29)30-21/h3-8,26-27H,1-2H3

InChI key

FRPHFZCDPYBUAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

pH indicator

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kerstin Müller et al.
Veterinary clinical pathology, 39(1), 79-82 (2009-08-26)
In reptile medicine, plasma chemistry analysis is widely used for the evaluation of an individual's health status. The standard method for the determination of plasma albumin concentration is protein electrophoresis combined with the determination of total protein concentration, but the
Kohji Hirata et al.
Research communications in molecular pathology and pharmacology, 117-118, 115-124 (2008-04-23)
Administration of antitumor drugs such as 5-fluorouracil and methotrexate to rats causes intestinal injury. The intestinal injury was reported to be prevented by coadministration of prostaglandin E1 analogues to rats. IEC-6 cell is a useful in vitro model to study
Carolyn Cray et al.
Journal of avian medicine and surgery, 25(1), 14-22 (2011-06-11)
The purpose of this study was to examine the validity of albumin determinations in penguin plasma by the bromcresol green (BCG) method and the gold standard of protein electrophoresis (EPH). Plasma from 96 clinically normal and abnormal penguins (Spheniscus species)
Kai Yu et al.
Enzyme and microbial technology, 49(3), 272-276 (2011-11-25)
A pH-sensitive colorimetric assay has been established to quantitatively measure glutamate decarboxylase (GAD) activity in bacterial cell extracts using a microplate format. GAD catalyzes the irreversible α-decarboxylation of L-glutamate to γ-aminobutyrate. The assay is based on the color change of
Yu-Chang Chang et al.
Sensors (Basel, Switzerland), 11(4), 4060-4072 (2011-12-14)
A meso-structured Al-MCM-41 material was impregnated with bromocresol green (BG) dye and then incorporated into a UV-Vis DRA spectroscopic instrument for the online detection of ammonia gas. The absorption response of the Al-MCM-41/BG ammonia sensing material was very sensitive at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service