Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40-2004

Sigma-Aldrich

Thymol blue solution

pH 1.2-2.8, pH 8.0-9.6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O5S
CAS Number:
Molecular Weight:
466.59
Beilstein/REAXYS Number:
368036
MDL number:
UNSPSC Code:
12171502
PubChem Substance ID:
Pricing and availability is not currently available.

form

liquid

availability

available only in Japan

dilution

(for analytical testing)

pH

1.2-2.8
8.0-9.6

SMILES string

CC(C)c1cc(c(C)cc1O)C(=C2\C=C(C(C)C)C(=O)C=C2C)\c3ccccc3S(O)(=O)=O

InChI

1S/C27H30O5S/c1-15(2)19-13-22(17(5)11-24(19)28)27(21-9-7-8-10-26(21)33(30,31)32-27)23-14-20(16(3)4)25(29)12-18(23)6/h7-16,28-29H,1-6H3

InChI key

PRZSXZWFJHEZBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

pH indicator

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Balderas-Hernández et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 66(1), 68-73 (2006-07-11)
This paper presents the results concerning the determination of the formation constants of the complexes between thymol blue, TB, and the metal ions Pb(II) or Cd(II). The experimental procedure was carried out in the presence of a nitrogen atmosphere at
N Gayathri et al.
Journal of AOAC International, 84(4), 1065-1069 (2001-08-15)
A simple and sensitive spectrophotometric method was developed for the determination of trace amounts of sulfur dioxide. The method is based on the reaction of SO2 with a known excess of ICI as the oxidant. The unreacted ICI iodinates thymol
Yong-Doo Park et al.
Journal of protein chemistry, 22(5), 473-480 (2003-12-24)
Sensitive assay methods for tyrosinase are essential not only for the understanding the process of pigment production but also for the development of effective inhibitors of tyrosinase. To develop an efficient assay method, we applied thymol blue to reaction mixtures.
Kanakapura Basavaiah et al.
Anais da Academia Brasileira de Ciencias, 80(2), 253-262 (2008-05-29)
Four sensitive and rapid methods for the determination of stavudine (STV) in bulk drug and in dosage forms were developed and optimized. In titrimetry, aqueous solution of STV was treated with a known excess of bromate-bromide in HCl medium followed
Shangjie Xu et al.
Macromolecular bioscience, 7(8), 968-974 (2007-08-01)
A simple general synthetic concept to build dendritic core-shell architectures with pH-labile linkers based on hyperbranched PEI cores and biocompatible PEG shells is presented. Using these dendritic core-shell architectures as nanocarriers, the encapsulation and transport of polar dyes of different

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service