Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37-0540

Sigma-Aldrich

Zinc stearate

CP

Synonym(s):

Stearic acid zinc salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)16COO]2Zn
CAS Number:
Molecular Weight:
632.33
Beilstein/REAXYS Number:
3919706
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade:
CP
form:
solid
Pricing and availability is not currently available.

grade

CP

form

solid

autoignition temp.

1454 °F

reaction suitability

reagent type: catalyst
core: zinc

availability

available only in Japan

mp

128-130 °C (lit.)

storage temp.

15-25°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)O[Zn]OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/2C18H36O2.Zn/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2

InChI key

XOOUIPVCVHRTMJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

356.0 °F - open cup

flash_point_c

180 °C - open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service