Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

35-0170

Sigma-Aldrich

p-Xylene

SAJ special grade, ≥99.0%

Synonym(s):

1,4-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Beilstein/REAXYS Number:
1901563
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
grade:
SAJ special grade
assay:
≥99.0%
bp:
138 °C (lit.)
vapor pressure:
9 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

SAJ special grade

vapor density

3.7 (vs air)

vapor pressure

9 mmHg ( 20 °C)

assay

≥99.0%

form

liquid

autoignition temp.

984 °F

expl. lim.

7 %

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.495 (lit.)

bp

138 °C (lit.)

mp

12-13 °C (lit.)

density

0.861 g/mL at 20 °C (lit.)

SMILES string

Cc1ccc(C)cc1

InChI

1S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6H,1-2H3

InChI key

URLKBWYHVLBVBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

80.6 °F

flash_point_c

27 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Venkatesha R Hathwar et al.
IUCrJ, 2(Pt 5), 563-574 (2015-08-26)
Rubrene is one of the most studied organic semiconductors to date due to its high charge carrier mobility which makes it a potentially applicable compound in modern electronic devices. Previous electronic device characterizations and first principles theoretical calculations assigned the
Francesc X Prenafeta-Boldú et al.
FEMS microbiology ecology, 80(3), 722-734 (2012-03-03)
The treatment of air contaminated with toluene, ethylbenzene, and p-xylene was assayed in three laboratory-scale biofilters, each consisting of two modules connected in series, packed with a pelletized organic fertilizer and inoculated with a toluene-degrading liquid enrichment culture. Biofilters were
G Gallastegui et al.
Journal of hazardous materials, 185(2-3), 1019-1026 (2010-10-30)
The influence of water irrigation on both the long-term and short-term performance of p-xylene biodegradation under several organic loading scenarios was investigated using an organic packing material composed of pelletised sawdust and pig manure. Process operation in a modular biofilter
A Vyskocil et al.
Toxicology and industrial health, 28(9), 796-819 (2011-11-09)
There is accumulating epidemiological evidence that exposure to some solvents, metals, asphyxiants and other substances in humans is associated with an increased risk of acquiring hearing loss. Furthermore, simultaneous and successive exposure to certain chemicals along with noise can increase
Martin Kuss-Petermann et al.
Journal of the American Chemical Society, 134(30), 12844-12854 (2012-07-20)
A homologous series of four molecules in which a phenol unit is linked covalently to a rhenium(I) tricarbonyl diimine photooxidant via a variable number of p-xylene spacers (n = 0-3) was synthesized and investigated. The species with a single p-xylene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service