Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33-0600

Sigma-Aldrich

(±)-α-Tocopherol

SAJ special grade, ≥96.0%

Synonym(s):

DL-all-rac-α-Tocopherol, Vitamin E

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
Beilstein/REAXYS Number:
94012
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ special grade

assay

≥96.0%

form

liquid

availability

available only in Japan

color

yellow

density

0.950 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)CCCC(C)CCCC(C)CCCC1(C)CCc2c(C)c(O)c(C)c(C)c2O1

InChI

1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3

InChI key

GVJHHUAWPYXKBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

507.2 °F

flash_point_c

264 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali A Albarhani et al.
Clinical biochemistry, 48(16-17), 1105-1112 (2015-04-30)
In widely used protocols for the collection and isolation of cord blood mononuclear cells, investigators are left with substantial volumes of diluted plasma which could be used for other measurements. The aim of this study was to ascertain the validity
Kirsten G Coupland et al.
Movement disorders : official journal of the Movement Disorder Society, 29(13), 1606-1614 (2014-01-01)
Parkinson's disease (PD) is a neurodegenerative disorder for which environmental factors influence disease risk and may act via an epigenetic mechanism. The microtubule-associated protein tau (MAPT) is a susceptibility gene for idiopathic PD. Methylation levels were determined by pyrosequencing of
Hannah Arem et al.
JAMA internal medicine, 175(6), 959-967 (2015-04-07)
The 2008 Physical Activity Guidelines for Americans recommended a minimum of 75 vigorous-intensity or 150 moderate-intensity minutes per week (7.5 metabolic-equivalent hours per week) of aerobic activity for substantial health benefit and suggested additional benefits by doing more than double
Sam B Weber et al.
Proceedings. Biological sciences, 279(1731), 1077-1084 (2011-09-23)
The effect of climate warming on the reproductive success of ectothermic animals is currently a subject of major conservation concern. However, for many threatened species, we still know surprisingly little about the extent of naturally occurring adaptive variation in heat-tolerance.
Siddharth Singh et al.
Clinical gastroenterology and hepatology : the official clinical practice journal of the American Gastroenterological Association, 13(4), 643-654 (2014-04-29)
Little is known about differences in rates of fibrosis progression between patients with nonalcoholic fatty liver (NAFL) vs nonalcoholic steatohepatitis (NASH). We conducted a systematic review and meta-analysis of all studies that assessed paired liver biopsy specimens to estimate the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service